missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin 3 Rabbit Anti-Human, Polyclonal, Abnova&Trade;
Rabbit Polyclonal Antibody
Brand: Abnova PAB1728.400uL
This item is not returnable.
View return policy
Description
ataxin 3 Polyclonal antibody specifically detects ataxin 3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ataxin 3 | |
| Polyclonal | |
| Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. |
|
| AT3/ATX3/JOS/MJD/MJD1/SCA3 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 29-59 at N-terminus of human ATXN3. | |
| RUO | |
| 4287 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| In PBS (0.09% sodium azide) | |
| Rabbit | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction