missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin 7 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-42657CL1
This item is not returnable.
View return policy
Description
Ataxin 7 Polyclonal antibody specifically detects Ataxin 7 in Human samples. It is validated for Western Blot,Immunoprecipitation (Negative)
Specifications
| Ataxin 7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6314 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Western Blot, Immunoprecipitation (Negative) | |
| CoraFluor 1 | |
| ADCAII, ataxin 7, FLJ17787, SCA7 | |
| The immunogen for this product maps to a region between residue 842 and 892 of human spinocerebellar ataxia type 7 using the numbering given in entry NP_000324.1 (GeneID 6314). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction