missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATCAY Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109423
This item is not returnable.
View return policy
Description
Immunogen sequence: MGTTEATLRM ENVDVKEEWQ DEDLPRPLPE ETGVELLGSP VEDTSSPPNT LNFNGAHRKR KTLVAPEINI SLDQSEGSLL SDDFLDTPDD LDINVDDIET PDETD.
This gene encodes a neuron-restricted protein that contains a CRAL-TRIO motif common to proteins that bind small lipophilic molecules. Mutations in this gene are associated with cerebellar ataxia, Cayman type.
Specifications
| ATCAY | |
| Polyclonal | |
| Unconjugated | |
| ATCAY | |
| 3322401A10Rik; ataxia cayman type protein; ataxia cerebellar Cayman type; ataxia, cerebellar, Cayman type; ataxia, cerebellar, Cayman type homolog (human); Atcay; ATCAY, caytaxin; BB077577; BNIP-2-homolgy; BNIP-2-homology; BNIP-H; Cayman ataxia; caytaxin; CLAC; hes; hesitant; ji; jittery; KIAA1872 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16467, 362826 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.78 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q1M168, Q8BHE3 | |
| ATCAY | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human ATCAY (NP_149053.1). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction