Learn More
Invitrogen™ ATE1 Recombinant Rabbit Monoclonal Antibody (24GB4720)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557343
 View more versions of this product
Product Code. 30275146
  • € 564.00 / 100µL
Description
ATE1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Arginyl-tRNA-protein transferase (ATE1), also designated arginyltransferase 1, belongs to the R-transferase family of proteins. In order for a protein to be degraded via the ubiquitin pathway, arginylation of the protein is required. ATE1 plays an important role in this process, as it is important for the posttranslational conjugation of arginine to the N-terminal aspartate-, glutamate- and possibly cystine-contaning substrates. ATE1 is a 518 amino acid protein. Alternative splicing results in two distinct isoforms. ATE1, which is found as a monomer, can localize to the cytoplasm and/or the nucleus.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATE1
Recombinant Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95260, Q9Z2A5
ATE1
A synthesized peptide derived from human ATE1 (aa 39-52).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4720
Unconjugated
ATE1
AI225793; arginine-tRNA-protein transferase 1; arginine-tRNA--protein transferase 1; arginyltransferase 1; arginyl-tRNA-protein transferase; arginyl-tRNA--protein transferase 1; ATE1; AW547406; R-transferase 1
Rabbit
Affinity chromatography
RUO
11101, 11907, 293526
-20°C