missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF3 Antibody (CL1685), Novus Biologicals™
Mouse Monoclonal Antibody
€ 369.00 - € 564.00
Specifications
| Antigen | ATF3 |
|---|---|
| Clone | CL1685 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407581
|
Novus Biologicals
NBP2-34489-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18008895
|
Novus Biologicals
NBP2-34489 |
0.1 mL |
€ 564.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATF3 Monoclonal specifically detects ATF3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ATF3 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Transcription Factors and Regulators | |
| P18847 | |
| 467 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MMLQHPGQVSASEVSASAIVPCLSPPGSLVFEDFANLTPFVKEELRFAIQNKHLCHRMSSALESVTVSDRPLGVSITKAEVAPEEDERKKRRRERNKIAAAKCRNKKKEKTEC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL1685 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3 | |
| ATF3 | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title