missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 14 publications
€ 280.00
Specifications
| Antigen | ATF3 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Immunohistochemistry-Frozen, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ATF3 Polyclonal specifically detects ATF3 in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
| ATF3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Feline | |
| activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3 | |
| ATF3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Immunohistochemistry-Frozen, Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 467 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MMLQHPGQVSASEVSASAIVPCLSPPGSLVFEDFANLTPFVKEELRFAIQNKHLCHRMSSALESVTVSDRPLGVSITKAEVAPEEDERKKRRRERNKIAAAKCRNKKKEKTEC | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title