missing translation for 'onlineSavingsMsg'
Learn More

ATF4 Antibody (9E1), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 343.00 - € 533.00

Specifications

Antigen ATF4
Clone 9E1
Dilution Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30030815
View Documents
Novus Biologicals
NBP3-26497-50ul
50 μL
€ 343.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30036879
View Documents
Novus Biologicals
NBP3-26497-100ul
100 μL
€ 533.00
100µL
Voir les produits de remplacement
Économisez jusqu'à 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATF4 Monoclonal antibody specifically detects ATF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Spécification

Spécification

ATF4
Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200
Monoclonal
Purified
RUO
Human
Activating transcription factor 4, activating transcription factor 4 (tax-responsive enhancer element B67), cAMP-dependent transcription factor ATF-4, cAMP-responsive element-binding protein 2, CREB-2DNA-binding protein TAXREB67, cyclic AMP-dependent transcription factor ATF-4, Cyclic AMP-responsive element-binding protein 2, TaxREB67, TAXREB67CREB2cAMP response element-binding protein 2, Tax-responsive enhancer element-binding protein 67, TXREB
A synthesized peptide derived from Human ATF4 [UniProt P18848]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
9E1
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Unconjugated
Rabbit
Lipid and Metabolism, Phospho Specific, Unfolded Protein Response, Unfolded Protein Response (UPR)
PBS, pH 7.4, 150mM NaCl and 50% glycerol
468
IgG
Affinity purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.