Learn More
ATF4 Monoclonal Antibody (OTI1H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | ATF4 |
|---|---|
| Clone | OTI1H9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17557414
|
Origene Technologies
TA812797S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17537414
|
Origene Technologies
TA812797 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
ATF4 Monoclonal antibody specifically detects ATF4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ATF4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Activating transcription factor 4; activating transcription factor 4 (tax-responsive enhancer element B67); activating transcription factor 4a; activating transcription factor 4b1 (tax-responsive enhancer element B67); activating transcription factor ATF-4; ATF 4; atf4; Atf-4; atf4 {ECO:0000312; ATF4 protein; atf4a; atf4b1; C/ATF; C/EBP-related ATF; cAMP response element-binding protein 2; cAMP-dependent transcription factor ATF-4; cAMP-responsive element-binding protein 2; CREB 2; CREB2; CREB-2; Cyclic AMP-dependent transcription factor ATF-4; cyclic AMP-responsive element-binding protein 2; DNA-binding protein TAXREB67; hypothetical protein LOC509107; OTTHUMP00000199130; rATF-4; RGD:621863}; tax-re; TaxREB67; TaxREB67 homolog; tax-responsive enhancer element B67; tax-responsive enhancer element-binding protein 67; Tax-responsive enhancer element-binding protein 67 homolog; TXREB; wu:fj01c08; wu:fk30b04; zgc:85951 | |
| Full length human recombinant protein of human Atf4 (NP_001666) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Atf4 |
| OTI1H9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P18848, Q06507, Q9ES19 | |
| 11911, 468, 79255 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.