missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF6 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76675F
This item is not returnable.
View return policy
Description
ATF6 Polyclonal antibody specifically detects ATF6 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ATF6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ATF6. The immunogen is located within amino acids 470 - 520 of ATF6. Amino Acid Squence: HRHEVERTKSRRMTNN | |
| 0.1 mL | |
| Lipid and Metabolism, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 22926 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction