missing translation for 'onlineSavingsMsg'
Learn More

ATF7IP Polyclonal Antibody, Invitrogen™

Product Code. 15732741 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15732741 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15732741 Supplier Invitrogen Supplier No. PA554811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SMEIDQGEKN EDETSADLVE TINENVIEDN KSENILENTD SMETDEIIPI LEKLAPSEDE LTCFSKTSLL PIDETNPDLE EKMESSFGSP SKQESSESLP KEAFLVLSDE EDISGEKDES EVISQNETCS P Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 69%.

ATF7IP is a multifunctional nuclear protein that associates with heterochromatin. It can act as a transcriptional coactivator or corepressor depending upon its binding partners.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF7IP
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Atf7ip
Gene Accession No. Q6VMQ6
Gene Alias 2610204M12Rik; activating transcription factor 7 interacting protein; activating transcription factor 7-interacting protein 1; AM; ATF7-interacting protein; ATF7IP; ATFa-associated modulator; ATF-interacting protein; ATF-IP; FLJ10688; hAM; mAM; MBD1-containing chromatin-associated factor 1; MCAF; MCAF1; p621
Gene Symbols Atf7ip
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATF7IP
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55729
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.