missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG14 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36445H
This item is not returnable.
View return policy
Description
ATG14 Polyclonal antibody specifically detects ATG14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| ATG14 | |
| Polyclonal | |
| Western Blot, ELISA | |
| ATG14 Autophagy Related 14 Homolog, ATG14 Autophagy Related 14 Homolog (S. Cerevisiae), ATG14L, Autophagy Related 14, Autophagy-Related Protein 14-Like Protein, Barkor, Beclin 1-Associated Autophagy-Related Key Regulator, Beclin 1-interacting protein, KIAA0831 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human ATG14. The immunogen is located within amino acids 270 - 320 of ATG14. Amino Acid Squence: VEEKKTTQGPDMEQSNPA | |
| 0.1 mL | |
| Autophagy, Cell Biology, Endocrinology, Signal Transduction | |
| 22863 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| HRP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction