missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG3/APG3 Antibody (OTI3H2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72140PCP
This item is not returnable.
View return policy
Description
ATG3/APG3 Monoclonal specifically detects ATG3/APG3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| ATG3/APG3 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| ATG3 | |
| Full length recombinant protein of human ATG3/APG3 (NP_071933) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
| Flow Cytometry | |
| OTI3H2 | |
| Flow Cytometry | |
| APG3, APG3 autophagy 3-like (S. cerevisiae), APG3L, APG3-LIKE, ATG3 autophagy related 3 homolog (S. cerevisiae), Autophagy-related protein 3, DKFZp564M1178, EC 6.3.2.-, FLJ22125,2610016C12Rik, hApg3, MGC15201, PC3-96, Protein PC3-96, ubiquitin-like-conjugating enzyme ATG3 | |
| Mouse | |
| Affinity Purified | |
| Autophagy, Cancer | |
| 64422.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction