missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-41307-0.025mg
This item is not returnable.
View return policy
Description
ATG4B Polyclonal specifically detects ATG4B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| ATG4B | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ATG4B | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human ATG4B. The immunogen is located within the last 50 amino acids of ATG4B. Amino Acid Squence: LDSSDVERLERFFDSED | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least two isoforms of ATG4B are known to exist; this antibody will detect only the larger isoform. ATG4B is predicted to not cross-react with other ATG4 proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| APG4 autophagy 4 homolog B, APG4 autophagy 4 homolog B (S. cerevisiae), APG4B, ATG4 autophagy related 4 homolog B (S. cerevisiae), AUTL1MGC1353, AUT-like 1 cysteine endopeptidase, Autophagin-1, Autophagy-related cysteine endopeptidase 1, Autophagy-related protein 4 homolog B, cysteine protease ATG4B, DKFZp586D1822, EC 3.4.22, EC 3.4.22.-, hAPG4B, KIAA0943 | |
| Rabbit | |
| 46 kDa | |
| 0.025 mg | |
| Autophagy, Cancer | |
| 23192 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction