missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4B Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41307JF646
This item is not returnable.
View return policy
Description
ATG4B Polyclonal antibody specifically detects ATG4B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ATG4B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| APG4 autophagy 4 homolog B, APG4 autophagy 4 homolog B (S. cerevisiae), APG4B, ATG4 autophagy related 4 homolog B (S. cerevisiae), AUTL1MGC1353, AUT-like 1 cysteine endopeptidase, Autophagin-1, Autophagy-related cysteine endopeptidase 1, Autophagy-related protein 4 homolog B, cysteine protease ATG4B, DKFZp586D1822, EC 3.4.22, EC 3.4.22.-, hAPG4B, KIAA0943 | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human ATG4B. The immunogen is located within the last 50 amino acids of ATG4B. Amino Acid Squence: LDSSDVERLERFFDSED | |
| 0.1 mL | |
| Autophagy, Cancer | |
| 23192 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction