missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4C Antibody (2E10H7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61691-0.025ml
This item is not returnable.
View return policy
Description
ATG4C Monoclonal specifically detects ATG4C in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ATG4C | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| APG4 autophagy 4 homolog C, APG4 autophagy 4 homolog C (S. cerevisiae), APG4°C, ATG4 autophagy related 4 homolog C (S. cerevisiae), AUT (S. cerevisiae)-like 1, cysteine endopeptidase; AUT-like 1, cysteineendopeptidase (S. cerevisiae), AUTL1, AUTL3APG4-C, AUT-like 1, cysteine endopeptidase, AUT-like 3 cysteine endopeptidase, Autophagin-3, Autophagy-related cysteine endopeptidase 3, Autophagy-related protein 4 homolog C, cysteine protease ATG4°C, EC 3.4.22, EC 3.4.22.-, FLJ14867 | |
| Mouse | |
| 52.5 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 2E10H7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ATG4°C | |
| Purified recombinant fragment of human ATG4°C (AA: 321-458) expressed in E. Coli. | |
| Protein G purified | |
| Autophagy, Cancer | |
| 84938 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction