missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4D Polyclonal Antibody

ATG4D Polyclonal Antibody
Brand: Invitrogen PA5142459
This item is not returnable.
View return policy
Description
ATG4D Polyclonal Antibody for IHC (P)
Specifications
| ATG4D | |
| Polyclonal | |
| Unconjugated | |
| Atg4d | |
| 9830134P12Rik; APG4 (ATG4) autophagy-related homolog D; APG4 autophagy 4 homolog D; APG4D; APG4-D; ATG4 autophagy related 4 homolog D; Atg4d; Atg4dl; Autl4; AUT-like 4 cysteine endopeptidase; AUT-like 4, cysteine endopeptidase; Autophagin-4; autophagy related 4D cysteine peptidase; autophagy related 4D, cysteine peptidase; autophagy-related 4D; autophagy-related cysteine endopeptidase 4; Autophagy-related protein 4 homolog D; cysteine protease ATG4D; Cysteine protease ATG4D, mitochondrial; cysteine protease involved in autophagy; cysteine protease involved in autophagy APG4-D | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 84971 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q86TL0 | |
| Atg4d | |
| Peptide with sequence CSQLAQPTLRLPRT (aa 444-457). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction