missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG5 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-53818IR
This item is not returnable.
View return policy
Description
ATG5 Polyclonal antibody specifically detects ATG5 in Human, Mouse, Rat, Porcine, Alligator, Bovine, Drosophila, Fish, Guinea Pig, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, Radioimmune Assays, Proximity Ligation Assay, Electron Microscopy, Gene Knock-Out, KnockDown
Specifications
| ATG5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Alligator | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry | |
| DyLight 755 | |
| APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25 | |
| This ATG5 Antibody was made to a synthetic peptide of an N-terminal region of the human ATG5 protein (within residues 1-50) [Swiss-Prot Q9H1Y0]. | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway | |
| 9474 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction