missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATG7 Monoclonal Antibody (29CT10.3.12)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA537466

Product Code. 17117058

  • € 510.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ATG7 Monoclonal Antibody for Western Blot

This gene was identified based on homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae APG7. In the yeast, the protein appears to be required for fusion of peroxisomal and vacuolar membranes. The protein shows homology to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATG7
Monoclonal
Conc. not determined
ascites with 0.09% sodium azide; pH 7.4
O95352
ATG7
purified His-tagged APG7 protein.
RUO
10533
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
29CT10.3.12
Unconjugated
ATG7
1810013K23Rik; Agp7; APG7L; APG7-LIKE; ATG12-activating enzyme E1 ATG7; Atg7; ATG7 autophagy related 7 homolog; Atg7l; autophagy related 7; autophagy related 7 homolog; autophagy-related 7; autophagy-related protein 7; DKFZp434N0735; Gm21553; GSA7; hAGP7; ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7
Mouse
100 μL
Primary
Human
Antibody
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.