missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517216
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
This gene was identified based on homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae APG7. In the yeast, the protein appears to be required for fusion of peroxisomal and vacuolar membranes. The protein shows homology to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes.
Specifications
| ATG7 | |
| Polyclonal | |
| Unconjugated | |
| ATG7 | |
| 1810013K23Rik; Agp7; APG7L; APG7-LIKE; ATG12-activating enzyme E1 ATG7; Atg7; ATG7 autophagy related 7 homolog; Atg7l; autophagy related 7; autophagy related 7 homolog; autophagy-related 7; autophagy-related protein 7; DKFZp434N0735; Gm21553; GSA7; hAGP7; ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10533, 312647, 74244 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 8 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O95352, Q641Y5, Q9D906 | |
| ATG7 | |
| Synthetic peptide corresponding to the amino terminus of Atg7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction