Learn More
ATG9A Antibody (Atg9 14F2 8B1), Alexa Fluor™ 405, Novus Biologicals™
Hamster Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50109AF405
Product Code. 18139067
  • € 448.00 / 0.10mL
Description
ATG9A Monoclonal specifically detects ATG9A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATG9A
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
ATG9A
Conserved C-terminal synthetic peptide of Human ATG9A sequence
0.1 mL
Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway
79065
Store at 4C in the dark.
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown
Atg9 14F2 8B1
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208
Hamster
Protein G purified
RUO
Primary
Human, Mouse, Rat