missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG9A Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26074V
This item is not returnable.
View return policy
Description
ATG9A Polyclonal antibody specifically detects ATG9A in Human samples. It is validated for Immunoprecipitation
Specifications
| ATG9A | |
| Polyclonal | |
| Immunoprecipitation | |
| APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208 | |
| Produced in rabbits immunized with E. coli-derived Human ATG9A fragment | |
| 0.1 mL | |
| Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| 79065 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction