missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG9A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572256
This item is not returnable.
View return policy
Description
ATG9A Polyclonal Antibody for Western Blot
Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). Apg9 plays a direct role in the formation of the cytoplasm to vacuole targeting and autophagic vesicles, possibly serving as a marker for a specialized compartment essential for these vesicle-mediated alternative targeting pathways.
Specifications
| ATG9A | |
| Polyclonal | |
| Unconjugated | |
| ATG9A | |
| Apg9; APG9 autophagy 9-like 1; Apg911; APG9A; APG9L1; APG9-like 1; ATG 9A; Atg9; ATG9 autophagy related 9 homolog A; ATG9 autophagy related 9 homolog A (S. cerevisiae); Atg9a; Atg9l1; AU019532; autophagy 9-like 1 protein; autophagy protein 9; autophagy related 9A; autophagy-related 9A; autophagy-related 9-like 1; autophagy-related protein 9; autophagy-related protein 9A; FLJ22169; mATG9; MGD3208; RGD1310450; zgc:158700 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100009663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.38 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| 0 | |
| ATG9A | |
| KLH conjugated synthetic peptide between 573-605 amino acids of DANRE atg9a. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction