Learn More
ATG9A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26276-1-AP-150UL
 View more versions of this product
Product Code. 16874785
  • € 410.00 / 150µL
Description
Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). Apg9 plays a direct role in the formation of the cytoplasm to vacuole targeting and autophagic vesicles, possibly serving as a marker for a specialized compartment essential for these vesicle-mediated alternative targeting pathways.
Specifications
Specifications
ATG9A
Polyclonal
Unconjugated
ATG9A
APG9 like 1, APG9L1, ATG9A, Autophagy related protein 9A, MGD3208
Rabbit
Antigen Affinity Chromatography
RUO
245860, 79065
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q68FE2, Q7Z3C6
ATG9A
ATG9A Fusion Protein Ag24212
150 μL
Primary
Human, Mouse