Learn More
ATG9A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26276-1-AP-150UL
This item is not returnable.
View return policy
Description
Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). Apg9 plays a direct role in the formation of the cytoplasm to vacuole targeting and autophagic vesicles, possibly serving as a marker for a specialized compartment essential for these vesicle-mediated alternative targeting pathways.Specifications
| ATG9A | |
| Polyclonal | |
| Unconjugated | |
| ATG9A | |
| APG9 like 1, APG9L1, ATG9A, Autophagy related protein 9A, MGD3208 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 245860, 79065 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q68FE2, Q7Z3C6 | |
| ATG9A | |
| ATG9A Fusion Protein Ag24212 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |