missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG9B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520998
This item is not returnable.
View return policy
Description
A suggested positive control is Hela cell lysate. PA5-20998 can be used with blocking peptide PEP-1112.
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG9B plays a role in autophagy and it's highly expressed in placenta and pituitary gland.
Specifications
| ATG9B | |
| Polyclonal | |
| Unconjugated | |
| ATG9B | |
| Apg912; Apg9l2; APG9-like 2; Apgdc2; ATG9; ATG9 autophagy related 9 homolog B; Atg9b; autophagy 9-like 2 protein; autophagy related 9B; autophagy-related protein 9B; endothelial nitric oxide synthase antisense; eONE; Gm574; nitric oxide synthase 3 antisense; nitric oxide synthase 3-overlapping antisense gene protein; NOS3AS; Protein sONE; RGD1560887; SONE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 213948, 285973, 499973 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q674R7, Q6EBV9 | |
| ATG9B | |
| A 17 amino acid synthetic peptide near the carboxy terminus of human ATG9B. The immunogen is located within amino acids 850-900 of ATG9B. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction