missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATHL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566213
This item is not returnable.
View return policy
Description
Immunogen sequence: LRGSLYYLLSA LPQPKAPGYI CHGLSPGGLS NGSREECYWG HVFWDQDLWM FPSILMFHPE AARAILEYRI RTLDGALENA QNLGYQGAKF A Highest antigen sequence identity to the following orthologs - mouse 84%, rat 85%.
PGGHG gene ontology annotations related to this gene include carbohydrate metabolic process.
Specifications
| ATHL1 | |
| Polyclonal | |
| Unconjugated | |
| PGGHG | |
| Acid trehalase-like protein 1; ATH1, acid trehalase-like 1; ATHL1; PGGHG; Protein-glucosylgalactosylhydroxylysine glucosidase; testicular tissue protein Li 25 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80162 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q32M88 | |
| PGGHG | |
| Recombinant Human ATHL1. Recombinant protein control fragment (Product #RP-106894). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction