missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATIC Antibody (OTI1D2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70220AF700
This item is not returnable.
View return policy
Description
ATIC Monoclonal specifically detects ATIC in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| ATIC | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ATIC | |
| Full length recombinant protein of human ATIC (NP_004035) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| OTI1D2 | |
| Western Blot, Flow Cytometry | |
| 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMPcyclohydrolase, 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotidetransformylase/inosinicase, AICAR, AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme, AICARFT, AICARFT/IMPCHASE, bifunctional purine biosynthesis protein PURH, IMPCHASE, phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase, PURHFLJ93545 | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 471.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction