missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATIC Antibody (OTI1D2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70220CL1
This item is not returnable.
View return policy
Description
ATIC Monoclonal antibody specifically detects ATIC in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| ATIC | |
| Monoclonal | |
| CoraFluor 1 | |
| 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMPcyclohydrolase, 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotidetransformylase/inosinicase, AICAR, AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme, AICARFT, AICARFT/IMPCHASE, bifunctional purine biosynthesis protein PURH, IMPCHASE, phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase, PURHFLJ93545 | |
| Full length human recombinant protein of human ATIC(NP_004035) produced in HEK293T cell. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 471 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Western Blot, CyTOF | |
| OTI1D2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction