missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATIC Monoclonal Antibody (OTI1D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504553
This item is not returnable.
View return policy
Description
ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.Specifications
| ATIC | |
| Monoclonal | |
| 0.71 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O35567, P31939, Q9CWJ9 | |
| Atic | |
| Full length human recombit protein of human ATIC produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| ATIC | |
| AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 108147, 471, 488513, 81643 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction