missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATIC Monoclonal Antibody (OTI2C10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504554S
This item is not returnable.
View return policy
Description
ATIC Monoclonal antibody specifically detects ATIC in Canine, Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ATIC | |
| Monoclonal | |
| 1.09 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O35567, P31939, Q9CWJ9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 108147, 471, 488513, 81643 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2C10 | |
| Unconjugated | |
| ATIC | |
| 153761_at; 2610509C24Rik; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase L homeolog; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AA536954; AICAR; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICARFT; AICARFT/IMPCHASE; atic; atic.L; AW212393; BEST:GH11240; Bifunctional purine biosynthesis protein PURH; Bifunctional purine biosynthesis protein PURH, N-terminally processed; CG11089; CG11089-PA; CG11089-PB; CT30991; Dmel\CG11089; Dmel_CG11089; epididymis secretory sperm binding protein Li 70p; HEL-S-70p; IMP cyclohydrolase; IMP synthase; IMPCHASE; Inosinicase; OK/SW-cl.86; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase; PURH; RE29555p; uncharacterized protein; XELAEV_18044849mg | |
| Full length human recombinant protein of human ATIC produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction