Learn More
ATIC Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI2C10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504554
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.Specifications
| ATIC | |
| Monoclonal | |
| Unconjugated | |
| ATIC | |
| AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 108147, 471, 488513, 81643 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2C10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O35567, P31939, Q9CWJ9 | |
| ATIC | |
| Full length human recombit protein of human ATIC produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.