missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atlantic Blue™ Anti-Human CD73 (AD2), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech AB-65162-100TESTS
This item is not returnable.
View return policy
Description
The CD73 antibody from Proteintech is a mouse monoclonal antibody generated with pre-b leukemia cell line of human CD73. This antibody recognizes human antigen. The CD73 antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.Specifications
| CD73 | |
| Monoclonal | |
| 5 μl/test | |
| CD73; E5NT; NT; NT5; NTE; eN; eNT | |
| Pre-B leukemia cell line | |
| 100 Tests | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| AD2 | |
| Atlantic Blue | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 4907 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction