missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Atlastin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29125
This item is not returnable.
View return policy
Description
Atlastin-3 Polyclonal antibody specifically detects Atlastin-3 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Atlastin-3 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25923 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| atlastin 3, atlastin GTPase 3, atlastin-3, DKFZP564J0863, EC 3.6.5.- | |
| between 491 and 541 | |
| 20 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction