missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATM Antibody, Alexa Fluor™ 594, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-271AF594
This item is not returnable.
View return policy
Description
ATM Polyclonal antibody specifically detects ATM in Human samples. It is validated for Western Blot
Specifications
| ATM | |
| Polyclonal | |
| Western Blot | |
| AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated, ataxia telangiectasia mutated (includes complementation groups A, C and D), ATC, ATD, ATDC, ATE, ATM serine/threonine kinase, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1 | |
| Region between residues 2550 and 2600 of human ATM (Ataxia Telangiectasia Mutated) | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, Modulation of DNA Pools, p53 Pathway, Protein Kinase, Tumor Suppressors | |
| 472 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction