missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATM, p Ser1981 Antibody (1C5), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 355.00 - € 568.00
Specifications
| Antigen | ATM |
|---|---|
| Clone | 1C5 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30031660
|
Novus Biologicals
NBP3-26430-50ul |
50 μL |
€ 355.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30026778
|
Novus Biologicals
NBP3-26430-100ul |
100 μL |
€ 568.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATM Monoclonal antibody specifically detects ATM in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
| ATM | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated, ataxia telangiectasia mutated (includes complementation groups A, C and D), ATC, ATD, ATDC, ATE, ATM serine/threonine kinase, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1 | |
| A synthesized peptide derived from Human ATM [p Ser1981] [UniProt Q13315] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1C5 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, Modulation of DNA Pools, p53 Pathway, Protein Kinase, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 472 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title