missing translation for 'onlineSavingsMsg'
Learn More

ATN-161 trifluoroacetate salt, MedChemExpress

Product Code. 16481897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481897

Brand: Medchem Express HY13535A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ATN-161 trifluoroacetate salt is a novel integrin α5β1 antagonist, which inhibits angiogenesis and growth of liver metastases in a murine model.

ATN-161 trifluoroacetate salt is a novel integrin α5β1 antagonist, which inhibits angiogenesis and growth of liver metastases in a murine model.

Specifications

Chemical Name or Material ATN-161 trifluoroacetate salt
CAS 904763-27-5
Color White
Purity Grade Notes Research
Molecular Formula C25H36F3N9O10S
Quantity 50 mg
Synonym ATN-161 TFA salt
Solubility Information H2O : 5 mg/mL (7.03 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 1 mg/mL (1.41 mM; Need ultrasonic)
SMILES FC(F)(C(O)=O)F.O=C(N)C[C@@H](C(N)=O)NC([C@H](CS)NC([C@H](CO)NC([C@H](CC1=CNC=N1)NC([C@H]2N(C(C)=O)CCC2)=O)=O)=O)=O
Molecular Weight (g/mol) 711.67
Formula Weight 711.67
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.