missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATOH1 Recombinant Rabbit Monoclonal Antibody (23GB4335), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | ATOH1 |
|---|---|
| Clone | 23GB4335 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088776
![]() |
Invitrogen™
MA552690 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087851
|
Invitrogen™
MA552691 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATOH1 Recombinant Monoclonal antibody specifically detects ATOH1 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| ATOH1 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92858 | |
| 474 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4335 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ATH1; Atoh1; atonal bHLH transcription factor 1; atonal homolog 1; atonal homolog 1 (Drosophila); atonal homolog bHLH transcription factor 1; BHLHA14; Class A basic helix-loop-helix protein 14; H MATH-1; Hath1; Helix-loop-helix protein hATH-1; helix-loop-helix protein mATH-1; Math1; MATH-1; Protein atonal homolog 1; RGD1565171 | |
| A synthesized peptide derived from human MATH1/HATH1 (1-51AA). | |
| Primary | |
| -20°C | |
| Atoh1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title