missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Epoxy Tetrahydrofuran Impurity, TRC

Product Code. 30136804
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.5mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136804 0.5 mg 0.5mg
30003647 5 mg 5mg
30010113 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136804 Supplier Toronto Research Chemicals Supplier No. TRCA7918600.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873950-19-7
Molecular Formula C26 H24 F N O5
Molecular Weight (g/mol) 449.47
Synonym 4-(4-Fluorophenyl)-2,4-dihydroxy-2-(1-methylethyl)-N,5-diphenyl-3,6-dioxabicyclo[3.1.0]hexane-1-carboxamide
IUPAC Name 4-(4-fluorophenyl)-2,4-dihydroxy-N,5-diphenyl-2-propan-2-yl-3,6-dioxabicyclo[3.1.0]hexane-1-carboxamide
SMILES CC(C)C1(O)OC(O)(c2ccc(F)cc2)C3(OC13C(=O)Nc4ccccc4)c5ccccc5

Specifications

Analyte or Component Names Atorvastatin Epoxy Tetrahydrofuran Impurity
Shipping Condition Room Temperature
InChI Formula InChI=1S/C26H24FNO5/c1-17(2)25(30)24(22(29)28-21-11-7-4-8-12-21)23(32-24,18-9-5-3-6-10-18)26(31,33-25)19-13-15-20(27)16-14-19/h3-17,30-31H,1-2H3,(H,28,29)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 0.5 mg
Formula Weight 449.1639
Chemical Name or Material Atorvastatin Epoxy Tetrahydrofuran Impurity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.