missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATOX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576204
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ATOX1 is a copper chaperone that plays a role in copper homeostasis by binding and transporting cytosolic copper to ATPase proteins in the trans-Golgi network for later incorporation to the ceruloplasmin. This protein also functions as an antioxidant against superoxide and hydrogen peroxide, and therefore, may play a significant role in carcinogenesis.
Specifications
| ATOX1 | |
| Polyclonal | |
| Unconjugated | |
| ATOX1 | |
| antioxidant 1 copper chaperone; ATOX1; ATX1; ATX1 antioxidant protein 1 homolog; copper transport protein ATOX1; HAH1; Metal transport protein ATX1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 475 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00244 | |
| ATOX1 | |
| Recombinant protein of human ATOX1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction