missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATOX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522973
This item is not returnable.
View return policy
Description
Suggested positive control: antigen standard for ATOX1 (transient overexpression lysate), HeLa nuclear extracts.
ATOX1 is a copper chaperone that plays a role in copper homeostasis by binding and transporting cytosolic copper to ATPase proteins in the trans-Golgi network for later incorporation to the ceruloplasmin. This protein also functions as an antioxidant against superoxide and hydrogen peroxide, and therefore, may play a significant role in carcinogenesis.
Specifications
| ATOX1 | |
| Polyclonal | |
| Unconjugated | |
| ATOX1 | |
| antioxidant 1 copper chaperone; ATOX1; ATX1; ATX1 antioxidant protein 1 homolog; copper transport protein ATOX1; HAH1; Metal transport protein ATX1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.1% sodium azide | |
| O00244 | |
| ATOX1 | |
| Full length human ATOX1 protein. | |
| RUO | |
| 475 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction