missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ATP Assay Kit (Colorimetric)

Product Code. 18672601 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18672601 96 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18672601 Supplier Novus Biologicals™ Supplier No. NBP324473
  • ONLINE EXCLUSIVE PRICE
    € 349.00
    € 329.70 / 1 sets
    Save € 19.30
    5% Off
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Assay Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage of components varies. See protocol for specific instructions.
Quantity 96 Tests
Species Non-species specific
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.