missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Citrate Lyase Antibody (OTI3G8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70073B
This item is not returnable.
View return policy
Description
ATP Citrate Lyase Monoclonal specifically detects ATP Citrate Lyase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ATP Citrate Lyase | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ACLY | |
| Full length recombinant protein of human ATP Citrate Lyase (NP_001087) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3G8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ACLEC 2.3.3.8, ATP citrate lyase, ATP citrate synthase, ATP-citrate (pro-S-)-lyase, ATP-citrate synthase, ATPCL, Citrate cleavage enzyme, CLATP | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific | |
| 47.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction