missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ATP synthase C mature Antibody (SR4230), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27824-50ul
This item is not returnable.
View return policy
Description
ATP synthase C mature Monoclonal antibody specifically detects ATP synthase C mature in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| ATP synthase C mature | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 516 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR4230 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9), ATP5A, ATP5G, ATPase protein 9, ATPase subunit 9, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9), isoform 1, mitochondrial, mitochondrial ATP synthase, subunit 9, mitochondrial ATP synthase, subunit C | |
| A synthesized peptide derived from human ATP synthase C mature (Uniprot #: P05496) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction