Learn More
ATP13A3 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-06070PE
Product Code. 18634988
  • € 430.00 / 0.10mL
Description
ATP13A3 Polyclonal antibody specifically detects ATP13A3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATP13A3
PE
PBS
Antigen and protein A Affinity-purified
Purified
Immunohistochemistry
Immunohistochemistry, Immunohistochemistry-Paraffin
AFURS1, ATPase family homolog up-regulated in senescence cells 1, ATPase type 13A3, DKFZp686K16189, FLJ90613, probable cation-transporting ATPase 13A3
79572