Learn More
Invitrogen™ ATP1A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120239
Description
Positive test controls include: U-87MG, OVCAR3, Mouse heart, Mouse lung, Mouse testis, Rat heart. The target is usually found in the following locations: Cell membrane, Membrane, Multi-pass membrane protein. Immunogen sequence: MGRGAGREYS PAATTAENGG GKKKQKEKEL DELKKEVAMD DHKLSLDELG RKYQVDLSKG LTNQRAQDVL.
Atp1a2 encodes an Na+/K+ ATPase, which is a membrane protein that is composed of two subunits - alpha and beta. The Na+/K+ ATPase pump maintains an essential electrochemical gradient within cells through the active transport of sodium (Na+) and potassium (K+) ions. The Atp1a2 encoded protein belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These ion gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. The ATPase enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta), and the catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. The Atp1a2 gene encodes an alpha 2 subunit. Mutations in the Atp1a2 gene result in familial basilar or hemiplegic migraines, and in a rare syndrome known as alternating hemiplegia of childhood. Diseases associated with ATP1A2 include Alternating Hemiplegia Of Childhood and Migraine, Familial Hemiplegic, 2.
Specifications
| ATP1A2 | |
| Polyclonal | |
| Unconjugated | |
| Atp1a2 | |
| Atp1a2; Atpa-3; ATPase Na+/K+ transporting alpha 2 polypeptide; ATPase Na+/K+ transporting subunit alpha 2; ATPase, Na+/K+ transporting, alpha 2 polypeptide; ATPase, Na+K+ transporting, alpha 2; AW060654; FHM2; KIAA0778; MHP2; mKIAA0778; Na(+)/K(+) ATPase alpha(+) subunit; Na(+)/K(+) ATPase alpha-2 subunit; Na+/K+ -ATPase alpha 2 subunit; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; RATATPA2; Sodium Potassium ATPase; Sodium pump subunit alpha-2; sodium/potassium-transporting ATPase alpha-2 chain; sodium/potassium-transporting ATPase subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24212, 477, 98660 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P06686, P50993, Q6PIE5 | |
| Atp1a2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human ATP1A2 (NP_000693.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.