missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP1A2 Recombinant Rabbit Monoclonal Antibody (5T3O4)
Shop All Thermo Scientific Products

Description
Positive Samples: Mouse heart, Mouse skeletal muscle, Rat heart Immunogen sequence: MGRGAGREYS PAATTAENGG GKKKQKEKEL DELKKEVAMD DHKLSLDELG RKYQVDLSKG LTNQRAQDVL ARDGPNALTP PPTTPEWVKF CRQLFGGFSI.
Atp1a2 encodes an Na+/K+ ATPase, which is a membrane protein that is composed of two subunits - alpha and beta. The Na+/K+ ATPase pump maintains an essential electrochemical gradient within cells through the active transport of sodium (Na+) and potassium (K+) ions. The Atp1a2 encoded protein belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These ion gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. The ATPase enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta), and the catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. The Atp1a2 gene encodes an alpha 2 subunit. Mutations in the Atp1a2 gene result in familial basilar or hemiplegic migraines, and in a rare syndrome known as alternating hemiplegia of childhood. Diseases associated with ATP1A2 include Alternating Hemiplegia Of Childhood and Migraine, Familial Hemiplegic, 2.
Specifications
Specifications
| Antigen | ATP1A2 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 5T3O4 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Atp1a2 |
| Gene Accession No. | P06686, Q6PIE5 |
| Gene Alias | Atp1a2; Atpa-3; ATPase Na+/K+ transporting alpha 2 polypeptide; ATPase Na+/K+ transporting subunit alpha 2; ATPase, Na+/K+ transporting, alpha 2 polypeptide; ATPase, Na+K+ transporting, alpha 2; AW060654; FHM2; KIAA0778; MHP2; mKIAA0778; Na(+)/K(+) ATPase alpha(+) subunit; Na(+)/K(+) ATPase alpha-2 subunit; Na+/K+ -ATPase alpha 2 subunit; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; RATATPA2; Sodium Potassium ATPase; Sodium pump subunit alpha-2; sodium/potassium-transporting ATPase alpha-2 chain; sodium/potassium-transporting ATPase subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?