missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP1A3 Polyclonal Antibody

Product Code. 16341245
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16341245 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16341245 Supplier Invitrogen™ Supplier No. PA584291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EVAMTEHKMS VEEVCRKYNT DCVQGLTHSK AQE Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

The sodium/potassium ATPase is an integral membrane enzyme found in all cells of higher organisms and is responsible for the ATP-dependent transport of sodium and potassium across the cell membrane. This membrane-bound enzyme is related to a number of other ATPases including sarcoplasmic and endoplasmic reticulum calcium ATPase (SERCA) and plasma membrane calcium ATPase (PMCA). The sodium/potassium ATPase consists of a large, multipass, transmembrane catalytic subunit, termed the alpha subunit, and an associated smaller glycoprotein, termed the beta subunit. Studies indicate that there are three isoforms of the alpha subunit (alpha 1, alpha 2, alpha 3) and two isoforms of the beta subunit (beta 1 and beta 2) encoded by two multigene families. The different isoforms of the sodium/potassium ATPase exhibit tissue-specific and developmental patterns of expression. The alpha 1 and beta mRNAs are present in all cell types examined, whereas the alpha 2 and alpha 3 mRNAs exhibit a more restricted pattern of cell-specific expression. The alpha-3 subunit has been found in neuronal and to skeletal and cardiac muscle, lung and stomach tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1A3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP1A3
Gene Accession No. P13637
Gene Alias AHC2; ATP1A1; Atp1a3; ATPa1a3; Atpa-2; ATPase Na+/K+ transporting subunit alpha 3; ATPase, Na+/K+ transporting, alpha 3 polypeptide; ATPase, Na+K+ transporting, alpha 3 subunit; CAPOS; DYT12; EC 3.6.3.9; Na(+)/K(+) ATPase alpha(III) subunit; Na(+)/K(+) ATPase alpha-3 subunit; Na+, K+ activated adenosine triphosphatase alpha subunit; Na+/K+ ATPase 3; Na+/K+ -ATPase alpha 3 subunit; RDP; Sodium Potassium ATPase; Sodium pump 3; sodium pump subunit alpha-3; sodium/potassium-transporting ATPase alpha-3 chain; sodium/potassium-transporting ATPase subunit alpha-3; sodium-potassium ATPase catalytic subunit alpha-3; sodium-potassium-ATPase, alpha 3 polypeptide
Gene Symbols ATP1A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP1A3. Recombinant protein control fragment (Product #RP-103256).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 478
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.