missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP1B2 Polyclonal Antibody

Product Code. 13280669
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13280669 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13280669 Supplier Invitrogen™ Supplier No. PA526279

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, mouse, rat and rabbit based on sequence homology.

This is the non-catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of Na+ and K+ ions across the plasma membrane. The exact function of the beta-2 subunit is not known. Mediates cell adhesion of neurons and astrocytes, and promotes neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1B2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ATP1B2
Gene Accession No. P14415
Gene Alias adhesion molecule in glia; adhesion molecule on glia; AMOG; ATP1B2; ATPase Na+/K+ transporting subunit beta 2; ATPase, Na+/K+ transporting, beta 2 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 2; ATPB2; Atpb-2; ATPB2S; glial cell adhesion molecule; Na, K-ATPase beta-2 polypeptide; Na,K-ATPase beta 2 subunit; Na+K+ ATPase beta 2 subunit; RATATPB2S; snoRNA MBI-85; Sodium Potassium ATPase; sodium pump subunit beta-2; sodium/potassium-dependent ATPase beta-2 subunit; sodium/potassium-dependent ATPase subunit beta-2; sodium/potassium-transporting ATPase beta-2 chain; sodium/potassium-transporting ATPase subunit beta-2; sodium-potassium ATPase subunit beta 2 (non-catalytic)
Gene Symbols ATP1B2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 115-141 amino acids from the central region of human ATP1B2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.