missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22338-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes the endogenous ATP1B2 protein in human brain. The bands between 45 kDa and 65 kDa represent the glycosylated forms of ATP1B2 in different levels.
This is the non-catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of Na+ and K+ ions across the plasma membrane. The exact function of the beta-2 subunit is not known. Mediates cell adhesion of neurons and astrocytes, and promotes neurite outgrowth.Specifications
| ATP1B2 | |
| Polyclonal | |
| Unconjugated | |
| ATP1B2 | |
| AMOG, ATP1B2, ATPase Na+/K+ beta 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11932, 24214, 482 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13638, P14231, P14415 | |
| ATP1B2 | |
| ATP1B2 Fusion Protein Ag17818 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction