missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP1B4 Antibody (4D5D4), Alexa Fluor™ 647 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05909AF647
This item is not returnable.
View return policy
Description
ATP1B4 Monoclonal antibody specifically detects ATP1B4 in Human samples. It is validated for ELISA
Specifications
| ATP1B4 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| ATPase, (Na+)/K+ transporting, beta 4 polypeptide, ATPase, Na+/K+ transporting, beta 4 polypeptide, K-ATPase beta-m subunit, K-ATPase subunit beta-m, protein ATP1B4, X, X/potassium-transporting ATPase subunit beta-m | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ATP1B4 (mature form, NP_036201.1; Tyr 132-Thr 357). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 4D5D4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 23439 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction