Learn More
ATP2A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22361-1-AP-20UL
 View more versions of this product
Product Code. 16871895
  • € 145.00 / 20µL
Description
ATP dependent calcium pumps are responsible, in part, for the maintece of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium (SERCA) ATPases. The SERCA1 gene is exclusively expressed in type II (fast) skeletal muscle. The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific form expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. The SERCA3 gene is not as well characterized and is found in non-muscle cells.
Specifications
Specifications
ATP2A1
Polyclonal
Unconjugated
ATP2A1
ATP2A, ATP2A1, Calcium pump 1, SERCA1, SR Ca(2+) ATPase 1
Rabbit
Antigen Affinity Chromatography
RUO
11937, 487
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14983, Q8R429
ATP2A1
ATP2A1 Fusion Protein Ag17944
20 μL
Primary
Human, Mouse